missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HAPLN4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant HAPLN4.
Brand: Abnova H00404037-A01.50uL
Additional Details : Weight : 0.00010kg
Description
Sequence: QRGRKKVVHVLEGESGSVVVQTAPGQVVSHRGGTIVLPCRYHYEAAAHGHDGVRLKWTKVVDPLAFTDVFVALGPQHRAFGSYRGRAELQGDGPGDASLVLRNVTSpecifications
HAPLN4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant HAPLN4. | |
HAPLN4 | |
BRAL2/KIAA1926 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_023002 | |
HAPLN4 | |
HAPLN4 (NP_075378, 30 a.a. ∽ 134 a.a) partial recombinant protein with GST tag. | |
RUO | |
404037 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only