missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Mouse anti-Human IgG Fc Secondary Antibody

Produktkod. 15358020
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Förpackningsstorlek:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
15358020 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15358020 Leverantör Invitrogen™ Leverantörsnummer MA517092

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Secondary Antibody

MA5-17092 targets human IgG (Fc Specific) in indirect ELISA, ICC, and WB applications and shows reactivity with Human samples. The MA5-17092 immunogen is purified fusion protein with human IgG(Fc Specific) tag.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Human IgG Fc
Användningsområden ELISA, Western Blot, Immunocytochemistry
Klassificering Monoclonal
Klona 4A10
Koncentration Conc. Not Determined
Konjugera Unconjugated
Formulering Ascites with 0.03% sodium azide
Värd art Mouse
Immunogen Purified fusion protein with human IgG(Fc Specific) tag.
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Secondary
Målarter Human
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Secondary Antibody
Form Liquid
Isotyp IgG1
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.