missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neisseria gonorrhoeae, Mouse, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against Neisseria gonorrhoeae.
Brand: Abnova MAB6185.100ug
Additional Details : Weight : 0.02000kg
Specifications
Neisseria gonorrhoeae | |
Monoclonal | |
Mouse monoclonal antibody raised against Neisseria gonorrhoeae. | |
In PBS, pH 7.4 | |
5766 and 8038 (W-II), 8660 (W-III)., Native Neisseria gonorrhoeae cells: Neisseria Reference Laboratory strains G-7, R-11and 7122 (W-I) | |
RUO | |
These antibodies do not cross-react with N. meningitidis, N. cinerea, N. lactamica, N. sicca, B. catarrhalis, E. coli, P. mirabilis, Gardnerella vaginialis, Group B Strep. or Chlamydia trachomatis. | |
Liquid |
ELISA, Immunofluorescence | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2b |