Learn More
IL16, Mouse, Clone: 210, Abnova™
Mouse monoclonal antibody raised against recombinant IL16.
Brand: Abnova MAB6290.500ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a pleiotropic cytokine that functions as a chemoattractant, a modulator of T cell activation, and an inhibitor of HIV replication. The signaling process of this cytokine is mediated by CD4. The product of this gene undergoes proteolytic processing, which is found to yield two functional proteins. The cytokine function is exclusively attributed to the secreted C-terminal peptide, while the N-terminal product may play a role in cell cycle control. Caspase 3 is reported to be involved in the proteolytic processing of this protein. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
IL16 | |
Monoclonal | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
IL16 | |
IL16 | |
Recombinant protein corresponding to human IL16. | |
RUO | |
3603 | |
Store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
|
IgG2a κ |
ELISA | |
210 | |
Mouse monoclonal antibody raised against recombinant IL16. | |
Lyophilized from 10mM PBS, pH 7.2 | |
FLJ16806/FLJ42735/FLJ44234/HsT19289/IL-16/LCF/prIL-16 | |
Mouse | |
500 μg | |
Primary | |
Human | |
Lyophilized |