Läs mer
42434, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MARCH5.
Brand: Abnova H00054708-A01.50uL
Beskrivning
MARCH5 is a ubiquitin ligase of the mitochondrial outer membrane that plays a role in the control of mitochondrial morphology by regulating mitofusin-2 (MFN2; MIM 608507) and DRP1 (DNM1L; MIM 603850) (Nakamura et al., 2006 [PubMed 16936636]).[supplied by OMIM
Sequence: MPDQALQQMLDRSCWVCFATDEDDRTAEWVRPCRCRGSTKWVHQACLQRWVDEKQRGNSTARVACPQCNAEYLIVFPKLGPVVYVLDLADRLISKSpecifikationer
| MARCH5. | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MARCH5. | |
| MARCH5 | |
| FLJ20445/MARCH-V/MITOL/RNF153 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_017824.4 | |
| MARCH5 | |
| 40973 (NP_060294, 1 a.a. ∽ 95 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 54708 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
For Research Use Only