missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I, Mouse, Clone: 34-1-2S, Abnova™

Mouse monoclonal antibody raised against MHC Class I.

Brand:  Abnova MAB11074.100ug

Additional Details : Weight : 0.02000kg

Product Code. 16089669

  • 3407.00 SEK / 100µg


Estimated Shipment
06-02-2020
Add to basket

Specifications

Specifications

MHC Class I
34-1-2S
Mouse monoclonal antibody raised against MHC Class I.
In aqueous buffer (0.09% sodium azide)
Mouse
IgG2a, kappa
100 ug
Store at 4°C.
Monoclonal
Flow Cytometry
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Mouse MHC Class I.
Affinity purification
RUO
Primary
Mouse
Videos
Documents