Läs mer
MPP1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Beskrivning
Palmitoylated membrane protein 1 is the prototype of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. Palmitoylated membrane protein 1 contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | MPP1 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Mouse polyclonal antibody raised against a full-length recombinant MPP1. |
| Formulering | 50% glycerol |
| Gen | MPP1 |
| Genaccessionsnr. | BC002392 |
| Gene Alias | AAG12/DXS552/DXS552E/EMP55/MRG1/PEMP |
| Gensymboler | MPP1 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.