missing translation for 'onlineSavingsMsg'
Läs mer

Mouse anti-Rat IgE Heavy Chain Secondary Antibody (MARE-1), Janelia Fluor 646, R&D Systems™

Produktkod. 30229329 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30229329 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30229329 Leverantör Novus Biologicals Leverantörsnummer NBP268394JF646
 Begränsad artikel
Beräknad leverans: 03-11-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

IgE Heavy Chain Monoclonal specifically detects IgE Heavy Chain in Rat samples. It is validated for ELISA, Western Blot.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen IgE Heavy Chain
Användningsområden ELISA, Western Blot
Klassificering Monoclonal
Klona MARE-1
Konjugera Janelia Fluor 646
Formulering 50mM Sodium Borate
Gene Alias Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon
Värd art Mouse
Immunogen Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins
Reningsmetod Protein G purified
Kvantitet 0.25 mL
Regulatorisk status RUO
Primär eller sekundär Secondary
Målarter Rat
Innehåll och lagring Store at 4°C in the dark.
Produkttyp Antibody
Form Purified
Isotyp IgG1
Produktlinje Novus
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.