Läs mer
Mouse anti-Rat IgG1 Heavy Chain Secondary Antibody (MARG1-2), Janelia Fluor 646, Novus Biologicals™
Handla allt Bio Techne ProdukterBeskrivning
IgG1 Heavy Chain Monoclonal specifically detects IgG1 Heavy Chain in Rat samples. It is validated for ELISA.
Specifikationer
Specifikationer
| Antigen | IgG1 Heavy Chain |
| Användningsområden | ELISA |
| Klassificering | Monoclonal |
| Klona | MARG1-2 |
| Konjugera | Janelia Fluor 646 |
| Formulering | 50mM Sodium Borate |
| Värd art | Mouse |
| Immunogen | Rat IR27 IgG1 myeloma protein |
| Reningsmetod | Affinity purified |
| Kvantitet | 0.25 mL |
| Visa mer |