Läs mer
SLC12A6, Mouse, Polyclonal Antibody, Abnova™
Beskrivning
This gene is a member of the K-Cl cotransporter (KCC) family. K-Cl cotransporters are integral membrane proteins that lower intracellular chloride concentrations below the electrochemical equilibrium potential. The proteins encoded by this gene are activated by cell swelling induced by hypotonic conditions. Alternate splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are associated with agenesis of the corpus callosum with peripheral neuropathy. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | SLC12A6 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Mouse polyclonal antibody raised against a partial recombinant SLC12A6. |
| Formulering | 50% glycerol |
| Gen | SLC12A6 |
| Genaccessionsnr. | NM_005135 |
| Gene Alias | ACCPN/DKFZp434D2135/KCC3/KCC3A/KCC3B |
| Gensymboler | SLC12A6 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.