Läs mer
SLU7, Mouse, Polyclonal Antibody, Abnova™
Beskrivning
Pre-mRNA splicing occurs in two sequential transesterification steps. The protein encoded by this gene is a splicing factor that has been found to be essential during the second catalytic step in the pre-mRNA splicing process. It associates with the spliceosome and contains a zinc knuckle motif that is found in other splicing factors and is involved in protein-nucleic acid and protein-protein interactions. [provided by RefSeq
Specifikationer
Specifikationer
| Antigen | SLU7 |
| Användningsområden | ELISA, Western Blot |
| Klassificering | Polyclonal |
| Konjugera | Unconjugated |
| Beskrivning | Mouse polyclonal antibody raised against a full-length recombinant SLU7. |
| Formulering | 50% glycerol |
| Gen | SLU7 |
| Genaccessionsnr. | BC010634 |
| Gene Alias | 9G8/MGC9280/hSlu7 |
| Gensymboler | SLU7 |
| Visa mer |
For Research Use Only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.