missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tag-72, Mouse, Clone: SPM536, Abnova™
Mouse monoclonal antibody raised against tag-72.
Brand: Abnova MAB12083.100ug
Additional Details : Weight : 0.02000kg
Specifications
tag-72 | |
Monoclonal | |
Unconjugated | |
Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
tag-72 | |
Mouse | |
Protein A/G purification | |
RUO | |
182875 | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 κ |
Immunohistochemistry (PFA fixed), Western Blot | |
SPM536 | |
Mouse monoclonal antibody raised against tag-72. | |
In PBS (0.05% BSA, 0.05% sodium azide) | |
tag-72 | |
Native purified human tag-72. | |
100 μg | |
Primary | |
Human | |
Liquid |