Learn More
TAZ, Mouse, Clone: 4A10-H3, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant TAZ.
Brand: Abnova H00006901-M01A.200uL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is expressed at high levels in cardiac and skeletal muscle. Mutations in this gene have been associated with a number of clinical disorders including Barth syndrome, dilated cardiomyopathy (DCM), hypertrophic DCM, endocardial fibroelastosis, and left ventricular noncompaction (LVNC). Multiple transcript variants encoding different isoforms have been described. A long form and a short form of each of these isoforms is produced; the short form lacks a hydrophobic leader sequence and may exist as a cytoplasmic protein rather than being membrane-bound. Other alternatively spliced transcripts have been described but the full-length nature of all these transcripts is not known. [provided by RefSeq
Sequence: MPLHVKWPFPAVPPLTWTLASSVVMGLVGTYSCFWTSEWAQAEAGPPGYPCPAGGILKLRHIWNLKLMRWTPAAADICFTKELHSHFFSLGKCVPVCRGDGVYQKGMDFILEKLNHGDWVHIFPEGIGRLIAECHLNPIILPLWHVGEPGDGDREMASGVGGLGLPLVPGCPAPPHVWPSVHCAAGMNDVLPNSPPYFPRFGQKITVLIGKPFSALPVLERLRAENKSAVEMRKALTDFIQEEFQHLKTQAEQLHNHLQPGRSpecifications
TAZ | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC011515 | |
TAZ | |
TAZ (AAH11515,1 a.a. ∽ 262 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
6901 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
4A10-H3 | |
Mouse monoclonal antibody raised against a full-length recombinant TAZ. | |
TAZ | |
BTHS/CMD3A/EFE/EFE2/FLJ27390/G4.5/LVNCX/Taz1/XAP-2 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |