Learn More
TGFB1, Mouse, Clone: 2C6, Abnova™
Mouse monoclonal antibody raised against native TGFB1.
Brand: Abnova MAB6319.500ug
Additional Details : Weight : 0.00010kg
Description
TGFB is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types. TGFB acts synergistically with TGFA (MIM 190170) in inducing transformation. It also acts as a negative autocrine growth factor. Dysregulation of TGFB activation and signaling may result in apoptosis. Many cells synthesize TGFB and almost all of them have specific receptors for this peptide. TGFB1, TGFB2 (MIM 190220), and TGFB3 (MIM 190230) all function through the same receptor signaling systems.[supplied by OMIM
Specifications
TGFB1 | |
Monoclonal | |
Unconjugated | |
Western Blot (50-500 ng/mL) The optimal working dilution should be determined by the end user. | |
TGFB1 | |
TGFB1 | |
Native purified human platelets TGFB1. | |
RUO | |
7040 | |
Human | |
Lyophilized |
ELISA, Immunohistochemistry, Inhibition Assays, Western Blot | |
2C6 | |
Mouse monoclonal antibody raised against native TGFB1. | |
Lyophilized from 10mM PBS, pH 7.0 | |
CED/DPD1/TGFB/TGFbeta | |
Mouse | |
500 μg | |
Primary | |
This antibody reacts with the dimeric (25 KDa) and monomeric (12.5 KDa) forms of TGFB1 under both non-reducing and reducing conditions respectively. | |
Store at -20°C. Aliquot after reconstitution to avoid repeated freezing and thawing. |
|
IgG1 κ |