missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse Bad N-Terminus Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB850
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Bad Monoclonal specifically detects Bad in Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifikationer
| Bad | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BAD | |
| E. coli-derived recombinant mouse Bad | |
| 100 μg | |
| Primary | |
| Detects endogenous mouse Bad in Western blots. In Western blots, this antibody does not cross-react with human Bad. The antibody also detects a 32 kDa band in Western blots. | |
| Mouse | |
| Purified |
| Western Blot | |
| 64103 | |
| Western Blot 1 ug/mL | |
| BBC2, BBC6, bcl2 antagonist of cell death, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, Bcl-2-binding component 6, BCL2-binding component 6, BCL2-binding protein, Bcl2-L-8, BCL2L8, BCL2L8bcl2-L-8, Bcl-2-like protein 8, BCL-X/BCL-2 binding protein, Bcl-XL/Bcl-2-associated death promoter | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 572 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering