missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse Fc gamma RI/CD64 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB20741-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Fc gamma RI/CD64 Monoclonal specifically detects Fc gamma RI/CD64 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifikationer
| Fc gamma RI/CD64 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P26151 | |
| Fcgr1 | |
| NS0-derived recombinant mouse Fc gamma RI extracellular domain Glu25-Pro297 Accession # P26151 | |
| 25 μg | |
| Primary | |
| Detects mouse Fc gamma RI in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Fc gamma RIA, rhFc gamma RIIB, or rhFc gamma RIIIB is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 290322 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD64, CD64 antigen, CD64a, CD64b, Fc fragment of IgG, high affinity Ia, receptor (CD64), FCG1, Fc-gamma receptor I B2, Fc-gamma RI, Fc-gamma RIA, FcgammaRIa, FCGR1, FCGR1B, FcgRI, FcgRIA, FCRI, FcRIA, FLJ18345, high affinity Ia, receptor for (CD64), high affinity immunoglobulin gamma Fc receptor I, IgG Fc receptor I | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2209 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering