missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse FGF-4 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB58461-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
FGF-4 Monoclonal specifically detects FGF-4 in Mouse samples. It is validated for Western Blot.
Specifikationer
| FGF-4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P11403 | |
| FGF4 | |
| E. coli-derived recombinant mouse FGF-4 Ala30-Leu202 Accession # P11403 | |
| 25 μg | |
| Primary | |
| Detects mouse FGF-4 in direct ELISAs and Western blots. In Western blots, 100% cross-reactivty with recombinant human (rh) FGF-4 and rhFGF-7 is observed. No cross-reactivity with rhFGF-3, -5, -9, -17, recombinant mouse FGF-6, -8b, -8c, or -10 is observed. | |
| Mouse | |
| Purified |
| Western Blot | |
| 678430 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF4, fibroblast growth factor 4, HBGF-4, HBGF-4Transforming protein KS3, heparin secretory transforming protein 1, Heparin secretory-transforming protein 1, Heparin-binding growth factor 4, HST-1, HST-1HSTF-1, HSTF1fibroblast growth factor 4 splice isoform, HSTFGF-4, human stomach cancer, transforming factor from FGF-related oncogene, kaposi sarcoma oncogene, KFGF, K-FGF, KS3, oncogene HST | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2249 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering