Läs mer
R&D Systems™ Mouse GDF-7/BMP-12 (NP_001299805.1) VersaClone cDNA
Specifikationer
Specifikationer
| Gen | GDF-7/BMP-12 |
| Tillträdesnummer | NP_001299805.1 |
| Gen-ID (Entrez) | 151449 |
| Gene Alias | BMP12, GDF-7, growth differentiation factor 7, growth/differentiation factor 7 |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Produktlinje | R&D Systems |
| Kvantitet | 10 μg |