Läs mer
Mouse HB-EGF Antibody, R&D Systems™
Rat Monoclonal Antibody
4480.00 SEK
Specifikationer
| Antigen | HB-EGF |
|---|---|
| Klona | 891808R |
| Utspädning | ELISA |
| Användningsområden | ELISA |
| Klassificering | Monoclonal |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
18734698
![]() |
R&D Systems
MAB82391-100 |
100 μg |
4480.00 SEK
100µg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
Beskrivning
HB-EGF Monoclonal specifically detects HB-EGF in Mouse samples. It is validated for ELISA.Specifikationer
| HB-EGF | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| Q06186 | |
| 1839 | |
| IgG2a | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 891808R | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor), Dtr, DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor), Dts, DTSF, HBEGF, Hegfl, heparin-binding EGF-like growth factor, heparin-binding epidermal growth factor, proheparin-binding EGF-like growth factor | |
| E. coli-derived recombinant mouse HB-EGF Asp63-Leu148 Accession # Q06186 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects mouse HB-EGF in direct ELISAs. |