missing translation for 'onlineSavingsMsg'
Läs mer

Proteintech™ Mouse IFN-gamma ELISA Kit

Produktkod. 16841596 Handla allt Proteintech Produkter
Klicka för att se tillgängliga alternativ
Målarter:
Mus
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 16841596

Brand: Proteintech™ KE1000196T

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

KE100001 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The IFN-γ ELISA kit is to be used to detect and quantify protein levels of endogenous IFN-γ.

  • The assay recognizes mouse IFN-γ. An antibody specific for IFN-γ has been pre-coated onto the microwells. The IFN-γ protein in samples is captured by the coated antibody after incubation.
  • Following extensive washing, another antibody of biotinylated specific for IFN-γ is added to detect the captured IFN-γ protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450nm with the correction wavelength set at 630 nm.
  • The IFNs were originally discovered as agents that interfere with viral replication. Initially, they were classified by the secreting cell type but are now classified into type I and type II according to receptor specificity and sequence homology.
  • Interferon gamma (IFN-γ) is a soluble cytokine that is the only member of the type II class of interferons. It is secreted by Th1 cells, cytotoxic T cells and NK cells.
  • The cytokine is associated with antiviral, immunoregulatory and anti-tumor properties and is a potent activator of macrophages. It plays crucial roles in pathogen clearance. Aberrant Ifng expression is associated with a number of autoinflammatory and autoimmune diseases.
  • It has been identified in many studies as a biomarker for pleural tuberculosis (TB). Mutations in this gene are associated with aplastic anemia.

Specifikationer

Tillträdesnummer NP_032363
Analysintervall 31,25 till 2000 pg/ml
Analyskänslighet 5.5 pg/mL
Certifikat och efterlevnad ISO 13485
Detektionsmetod Smörgås ELISA , dubbel antikropp
Immunoassay Kit Format Överdragen ELISA Utrustning
Provtyp Plasma, Serum
Substrat DAB
Målarter Mus
Gen-ID (Entrez) 15978
Värd art Mus
Immunoassay Kit Grade Fast
Interassay CV <10%
Intraassay CV <10%
Frakt skick Blå is
Förvaringskrav -20 °C till +4 °C
Total analystid 4.5 hr.
UniProt ID P01580
Visa mer Visa mindre
compliance-icons compliance-icons
Produktbeteckning
  • 96TEST Mouse IFN-gamma ELISA Kit
Signalord
  • Varning
Riskkategorier
  • Allvarlig ögonskada/ögonirritation Kategorija 1
  • Allvarlig ögonskada/ögonirritation Kategorija 2
  • Frätande eller irriterande verkan på hud Kategori 2
  • Specifik organtoxicitet Kategorija 3
Faroangivelser
  • H302-Skadligt vid förtäring.
  • H315-Irriterar huden.
  • H318-Orsakar allvarliga ögonskador.
  • H319-Orsakar allvarlig ögonirritation.
  • H332-Skadligt vid inandning.
  • H335-Kan orsaka irritation i luftvägarna.
Skyddsangivelse
  • P201-Inhämta särskilda instruktioner före användning.
  • P202-Använd inte produkten innan du har läst och förstått säkerhetsanvisningarna.
  • P234-Förvaras endast i originalbehållaren.
  • P264-Tvätta grundligt efter användning.
  • P270-Ät inte, drick inte och rök inte när du använder produkten.
  • P273-Undvik utsläpp till miljön.
  • P280-Använd skyddshandskar/skyddskläder/ögonskydd/ansiktsskydd.
  • P301+P310-VID FÖRTÄRING: Kontakta genast GIFTINFORMATIONSCENTRALEN/läkare/
  • P301+P312-VID FÖRTÄRING: Kontakta GIFTINFORMATIONSCENTRAL / läkare / /om du mår dåligt.
  • P302+P352-VID HUDKONTAKT: Tvätta med mycket vatten/.
  • P304+P340-VID INANDNING: Flytta personen till frisk luft och se till att andningen underlättas.
  • P308+P313-Vid exponering eller misstanke om exponering Sök läkarhjälp.
  • P330-Skölj munnen.
  • P362-Ta av nedstänkta kläder.
  • P405-Förvaras inlåst.
  • P501b-Innehållet/behållaren lämnas till I enlighet med lokala / regionala / nationella / internationella bestämmelser (ska anges).
Kompletterande information
  • MIXTURE LIST-Innehåller: Sulphuric Acid, Tetramethylbenzidine
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.