missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IgG Isotype Control, FITC, Polyclonal, Abnova™
Brand: Abnova PAB19457.500ug
Additional Details : Weight : 0.00010kg
Specifications
IgG Isotype Control | |
Polyclonal | |
FITC | |
Flow Cytometry (≤ 1 ug/106 cells) ELISA (1:500-1:5000) The optimal working dilution should be determined by the end user. | |
Mouse | |
Primary | |
Liquid |
ELISA, Flow Cytometry | |
0.5 mg/mL | |
Mouse IgG raised from normal mouse serum. | |
In PBS (0.09% sodium azide) | |
500 μg | |
Store at 4°C. Do not freeze. | |
IgG |
For Research Use Only