Learn More
Mouse IgG2b kappa Isotype Control (eBMG2b), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Isotype Control
1400.00 SEK - 3630.00 SEK
Specifications
Antigen | Mouse IgG2b kappa |
---|---|
Clone | eBMG2b |
Applications | Flow Cytometry |
Conjugate | Super Bright 436 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15835533
|
Affymetrix eBioscience
62-4732-82 |
100 μg |
3630.00 SEK
100µg |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
15825533
|
Invitrogen
62-4732-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The monoclonal mouse IgG2b is useful as an isotype control immunoglobulin. Applications Reported: This eBMG2b antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBMG2b antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This should be used at the same concentration as the experimental antibody. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer for more information. Excitation: 405 nm; Emission: 436 nm; Laser: Violet Laser Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Flow Cytometry.Specifications
Mouse IgG2b kappa | |
Flow Cytometry | |
Liquid | |
RUO | |
IgG2b κ | |
Affinity chromatography |
eBMG2b | |
Super Bright 436 | |
Mouse | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
Isotype Control (Primary) |
For Research Use Only