missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Mouse IL-9 Uncoated ELISA Kit

Produktkod. 15511207
Klicka för att se tillgängliga alternativ
Kvantitet:
10 x 96 tester
20 x 96 tester
Förpackningsstorlek:
Paket med 10
Paket med 20
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 15511207

Brand: Invitrogen™ 88809288

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

ELISA

The Mouse Interleukin 9 (IL-9) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-9. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.

Interleukin 9 (IL-9) is produced by activated T helper 2 (Th2) lymphocytes. IL-9 regulates many cell types involved in classical Th2-associated asthma responses, including B and T lymphocytes, mast cells, eosinophils, and epithelial cells. The mouse IL-9 cDNA encodes a 144 amino acid precursor protein with an 18 amino acid signal peptide that is cleaved to form the mature cysteine-rich protein with a molecular mass of 14 kDa. Native mouse IL-9 is heavily glycosylated. Recently, TGF beta has been reported to play an important role in the differentiation of IL-17-producing T helper cells and inducible regulatory T cells by reprogramming Th2 cells to secrete IL-9, thereby driving the differentiation of Th9 cells directly.
TRUSTED_SUSTAINABILITY

Specifikationer

Tillträdesnummer P15247
Analysintervall 32 till 4 000 pg/ml
Analyskänslighet 32 pg/mL
Innehåll och lagring Capture Antibody: Pre-titrated, purified antibody
Detection Antibody: Pre-titrated, biotin-conjugated antibody
Standard: Recombinant protein for generating standard curve and calibrating samples
Coating Buffer: 10X PBS ELISA Coating Buffer
Assay Diluent: 5X Concentrated
Detection Enzyme: Pre-titrated Avidin-HRP
Substrate Solution: Tetramethylbenzidine (TMB) Solution
Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards.
96-well Plates: Corning Costar flat-bottom plates (included with Cat. Nos. ending in -22, -76, and -86)
Konjugera HRP
Immunoassay Kit Format Uncoated ELISA Kit
Produkttyp ELISA
Provtyp Serum, Supernatant
Målarter Mus
För användning med (applikation) ELISA
För användning med (utrustning) Kolorimetrisk mikroplattläsare
Gene Alias cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40cytokine P40; homolog of mouse T cell and mast cell growth factor 40; HP40; IL9; IL-9; ILN; Interleukin; interleukin 9; Interleukin9; interleukin-9; P40; p40 cytokine; p40 T-cell and mast cell growth factor; T-cell growth factor P40; IL-2
Gen-ID (Entrez) 16198
Gensymbol IL-2
Kit Innehåll Capture Antibody: Förtitrerad, Renad antikropp, Detektionsantikropp: Förtitrerad, Biotinkonjugerad antikropp, Standard: Rekombinant protein för generering av standardkurvor och kalibrering av prover, Beläggningsbuffert: 10X PBS ELISA Beläggningsbuffert, Analysspädningsmedel: 5X koncentrerad, Detektionsenzym: Förtitrerad Streptavidin-HRP, Substratlösning: Tetrametylbensidin (TMB)-lösning, Analyscertifikat: Lotspecifika instruktioner för spädning av antikroppar och standarder, 96-brunnars platta: 96-brunnars platta med hög affinitet (ingår med produktkatalognummer som slutar på suffixen -22, -76, -86)
Etikett eller färgämne Biotin
Proteinaliaser IL-9, Cytokine P40, T-cell growth factor P40
Kvantitet 10 x 96 tester
Regulatorisk status RUO
Provvolym Serum 100μ L, Supernatant 100μ L
Förvaringskrav 2 °C till 8 °C
Synonym Interleukin-9, T-cellstillväxtfaktor P40
Mål IL-9
Testtid 1 tim 45 min
Total analystid 24 timmar 30 minuter
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.