Learn More
Novus Biologicals™ Mouse LIPC ELISA Kit (Colorimetric)
Mouse LIPC ELISA Kit (Colorimetric) measures LIPC in Serum, plasma, cell supernatant, and other biological fluids.
Brand: Novus Biologicals™ NBP3-00448
This item is not returnable.
View return policy
Specifications
0.39 to 25 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Human LIPC in samples. No significant cross-reactivity or interference between Human LIPC and analogues was observed. | |
ELISA | |
LIPC | |
Storage of components varies. See protocol for specific instructions. | |
LIPC |
0.23 ng/mL | |
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum | |
Mouse | |
3990 | |
1 Kit | |
EC 3.1.1, EC 3.1.1.3, Hepatic lipase, hepatic triacylglycerol lipase, HLHDLCQ12, HTGL, Lipase member C, lipase, hepatic, LIPH |