Learn More
Novus Biologicals™ Mouse LIPC ELISA Kit (Colorimetric)
Mouse LIPC ELISA Kit (Colorimetric) measures LIPC in Serum, plasma, cell supernatant, and other biological fluids.
Brand:  Novus Biologicals™ NBP3-00448
Product Code. 18322284
  • 7544.08 SEK / 1 set
Explore more special offers
Specifications
0.39 to 25 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Human LIPC in samples. No significant cross-reactivity or interference between Human LIPC and analogues was observed.
ELISA
LIPC
Storage of components varies. See protocol for specific instructions.
LIPC
0.23 ng/mL
Cell Culture Supernatant, Other Biological Fluids, Plasma, Serum
Mouse
3990
1 Kit
EC 3.1.1, EC 3.1.1.3, Hepatic lipase, hepatic triacylglycerol lipase, HLHDLCQ12, HTGL, Lipase member C, lipase, hepatic, LIPH