Learn More
Mouse pancreas tissue lysate - total protein, 1 mg
Product Code. 32152954 Shop All Abcam Products
Change view
Click to view available options
Unit Size:
1mg
Product Code. Unit Size
32152954 1mg
1 options
Product Code. 32152954 Supplier Abcam Supplier No. ab293631mg
 Restricted Item
Beräknad leverans: 30-11-2026
Mouse pancreas cell/tissue lysate.
  • Species: Mouse
  • Tissue: pancreas
  • Form: Liquid
Specifications
For Use With (Application) Western Blot