missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse Ret Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB482
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Ret Monoclonal specifically detects Ret in Mouse samples. It is validated for Western Blot.
Specifikationer
| Ret | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDHF12MTC1, CDHR16, EC 2.7.10, Hirschsprung disease 1, Hirschsprung disease), MEN2A, MEN2B, multiple endocrine neoplasia and medullary thyroid carcinoma 1, ret proto-oncogene, RET transforming sequence | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5979 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 64623 | |
| Western Blot 1 ug/mL | |
| P35546 | |
| RET | |
| Mouse myeloma cell line NS0-derived recombinant mouse Ret Leu29-Arg637 Accession # P35546 | |
| 500 μg | |
| Primary | |
| Detects mouse Ret in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 10-50% cross-reactivity with recombinant human Ret. | |
| Mouse | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering