Läs mer
R&D Systems™ Mouse Serpin E1/PAI-1 (NP_032897.2) VersaClone cDNA
Specifikationer
Specifikationer
| Gen | Serpin E1/PAI-1 |
| Tillträdesnummer | NP_032897.2 |
| Koncentration | 10 μg (at 0.2 μg/μL) |
| Formulering | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Infoga typ | 1222 bp |
| Gen-ID (Entrez) | 5054 |
| Gene Alias | Endothelial plasminogen activator inhibitor, Nexin, PAI1, PAI-1, PAI1PAI-1, PAISerpin E1, PLANH1, PLANH1plasminogen activator inhibitor 1, serine (or cysteine) proteinase inhibitor, clade E (nexin, plasminogenactivator inhibitor type 1), member 1, serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitortype 1), member 1 |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Art | Mouse |
| Kvantitet | 10 μg |