missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse Themis Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC6816R-100UG
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Themis Monoclonal specifically detects Themis in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifikationer
| Themis | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8BGW0 | |
| THEMIS | |
| E. coli-derived recombinant mouse Themis Lys122-Lys237 Accession # Q8BGW0 | |
| RUO | |
| 387357 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 719945 | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| bA325O24.3, bA325O24.4, C6orf190, C6orf207, FLJ40584, GASP, GRB2-associated protein, MGC163388, protein THEMIS, signaling phosphoprotein specific for T cells, SPOT, thymocyte selection associated, thymocyte selection pathway associated, thymocyte-expressed molecule involved in selection, TSEPA | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Themis in direct ELISAs and Western blots. In Western blots, 100% cross-reactivity with recombinant human (rh) Themis (aa 2-282) is observed under non-reducing conditions, and approximately 10% cross-reactivity with rhThemis (aa 2-282) is observed under reducing conditions. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only (RUO)
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering