missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse TIM-1/KIM-1/HAVCR APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1817A
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TIM-1/KIM-1/HAVCR Monoclonal specifically detects TIM-1/KIM-1/HAVCR in Mouse samples. It is validated for Flow Cytometry.
Specifikationer
| TIM-1/KIM-1/HAVCR | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD365, HAVCR1, HAVCR-1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26762 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 222414 | |
| Flow Cytometry 10 uL/10^6 cells | |
| NP_001160104 | |
| HAVCR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TIM-1/KIM-1/HAVCR Tyr22-Thr212 Accession # NP_001160104 | |
| 100 Tests | |
| Primary | |
| Detects mouse TIM-1/KIM-1/HAVCR in ELISAs and Western blots. In sandwich immunoassays, this antibody does not cross-react with recombinant human (rh) TIM-1, recombinant mouse (rm)TIM-2, or rmTIM-3. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering