Läs mer
R&D Systems™ Mouse TIM-1/KIM-1/HAVCR (NP_599009) VersaClone cDNA
Beskrivning
The Mouse TIM-1/KIM-1/HAVCR (NP_599009) VersaClone cDNA clone contains the complete ORF for TIM-1/KIM-1/HAVCR, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The TIM-1/KIM-1/HAVCR insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature
Specifikationer
Specifikationer
| Gene Alias | CD365, HAVcr-1, HAVCR1, HAVCRT cell immunoglobin domain and mucin domain protein 1, hepatitis A virus cellular receptor 1, Kidney injury molecule 1, KIM1, KIM-1, T-cell immunoglobulin and mucin domain-containing protein 1, TIM1, TIM-1TIM, TIM1TIMD-1, TIMD1T-cell membrane protein 1 |
| Innehåll och lagring | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Produktlinje | R&D Systems |
| Kvantitet | 10 μg |
For Reasearch Use Only; Not for Use in Humans