missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Mouse TM4SF1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7514
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TM4SF1/L6 Polyclonal specifically detects TM4SF1/L6 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifikationer
| TM4SF1/L6 | |
| Polyclonal | |
| Unconjugated | |
| Q64302 | |
| TM4SF1 | |
| E. coli-derived recombinant mouse TM4SF1 Ala116-Thr161 Accession # Q64302 | |
| 100 μg | |
| Primary | |
| Detects mouse TM4SF1 in direct ELISAs and Western blots. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| L6, M3s1, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4071 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering