Läs mer
Moxidectin EP L, TRC
Produktkod. 30466731
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30466731 5 mg
1 options
Produktkod. 30466731 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM7448105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (23Z)-Moxidectin
CAS 119718-45-5
Molekylformel C37 H53 N O8
Kvantitet 5 mg
Synonym Milbemycin B','5-O-demethyl-28-deoxy-25-(1,3-dimethyl-1-butenyl)-6,28-epoxy-23-(methoxyimino)-','[6R,23Z,25S(E)]- (9CI), Spiro[11,15-methano-2H,13H,17H-furo[4,3,2-pq][2,6]benzodioxacyclooctadecin-13,2'-[2H]pyran]','milbemycin B deriv., (23Z)-Moxidectin, Moxidectin Imp. L (EP)
InChI formel InChI=1S/C37H53NO8/c1-21(2)14-25(6)33-26(7)31(38-42-8)19-36(46-33)18-29-17-28(45-36)13-12-23(4)15-22(3)10-9-11-27-20-43-34-32(39)24(5)16-30(35(40)44-29)37(27,34)41/h9-12,14,16,21-22,26,28-30,32-34,39,41H,13,15,17-20H2,1-8H3/b10-9+,23-12+,25-14+,27-11+,38-31-/t22-,26-,28+,29-,30-,32+,33+,34+,36-,37+/m0/s1
LEDER CO\N=C/1\C[C@]2(C[C@@H]3C[C@@H](C\C=C(/C)\C[C@@H](C)\C=C\C=C\4/CO[C@@H]5[C@H](O)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2)O[C@@H]([C@H]1C)\C(=C\C(C)C)\C
Molekylvikt (g/mol) 639.82
Formel vikt 639.3771