Läs mer
Moxidectin O-TBS, TRC
Produktkod. 30472558
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30472558 100 mg
1 options
Produktkod. 30472558 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM744820100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Moxidectin O-TBS
CAS 1646166-08-6
Molekylformel C43 H67 N O8 Si
Kvantitet 100 mg
Synonym (6R,23E,25S)-5-O-Demethyl-28-deoxy-25-[(1E)-1,3-dimethyl-1-buten-1-yl]-5-O-[(1,1-dimethylethyl)dimethylsilyl]-6,28-epoxy-23-(methoxyimino)-milbemycin B
InChI formel InChI=1S/C43H67NO8Si/c1-26(2)19-29(5)37-31(7)36(44-47-11)24-42(51-37)23-34-22-33(50-42)18-17-28(4)20-27(3)15-14-16-32-25-48-39-38(52-53(12,13)41(8,9)10)30(6)21-35(40(45)49-34)43(32,39)46/h14-17,19,21,26-27,31,33-35,37-39,46H,18,20,22-25H2,1-13H3/b15-14+,28-17+,29-19+,32-16-,44-36+/t27-,31-,33+,34-,35-,37+,38+,39+,42-,43+/m0/s1
LEDER CO\N=C\1/C[C@]2(C[C@@H]3C[C@@H](C\C=C(/C)\C[C@@H](C)\C=C\C=C/4\CO[C@@H]5[C@H](O[Si](C)(C)C(C)(C)C)C(=C[C@@H](C(=O)O3)[C@]45O)C)O2)O[C@@H]([C@H]1C)\C(=C\C(C)C)\C
Molekylvikt (g/mol) 754.08
Formel vikt 753.4636