Läs mer
Moxifloxacin Hydrochloride, MedChemExpress
Produktkod. 16474067
Encompass_Preferred
Change view
Kvantitet:
50 mg
100 mg
500 mg
Förpackningsstorlek:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16474067 50 mg 50mg
16437916 100 mg -
16401946 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16474067 Leverantör Medchem Express Leverantörsnummer HY66011
MedChemExpress Moxifloxacin Hydrochloride (BAY 12-8039) is an oral 8-methoxyquinolone antimicrobial for use in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
Moxifloxacin Hydrochloride (BAY 12-8039) is an oral 8-methoxyquinolone antimicrobial for use in the treatment of acute bacterial sinusitis, acute bacterial exacerbations of chronic bronchitis, and community-acquired pneumonia.
Specifikationer
Kemiskt namn eller material Moxifloxacin Hydrochloride
Fysisk form Solid
CAS 186826-86-8
Färg Yellow
Kvantitet 50 mg
Anteckningar om renhetsgrad Research
Molekylformel C21H25ClFN3O4
För användning med (applikation) COVID-19-immunoregulation
Synonym BAY 12-8039
Löslighetsinformation DMSO : ≥ 31 mg/mL (70.79 mM) ∣H2O : 16.67 mg/mL (38.07 mM; Need ultrasonic)
LEDER O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N(C4)C[C@]5([H])[C@@]4([H])CCCN5)=C3OC)C1=O)O.Cl
Molekylvikt (g/mol) 437.89
Formel vikt 437.89
Procent renhet 99.51%