Learn More
Moxonidine, TRC
Product Code. 30133687
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133687 250 mg 250mg
30137265 50 mg 50mg
30041002 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133687 Supplier Toronto Research Chemicals Supplier No. TRCM745800250MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75438-57-2
Molecular Formula C9 H12 Cl N5 O
Molecular Weight (g/mol) 241.68
Synonym Moxonidine, 4-Chloro-N-(imidazolidin-2-ylidene)-6-methoxy-2-methylpyrimidin-5-amine, 5-Pyrimidinamine, 4-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-, 4-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-6-methoxy-2-methyl-5-pyrimidinamine, (4-Chloro-6-methoxy-2-methyl-pyrimidin-5-yl)-imidazolidin-2-ylidene-amine, 2-(6-Chloro-4-methoxy-2-methylpyrimidin-5-ylamino)-2-imidazoline, BDF 5895, BE 5895, Cynt, LY 326869, Lomox, Moxogamma, Moxon, Norcynt, Normoxocin, Nucynt, Physiotens
IUPAC Name N-(4-chloro-6-methoxy-2-methylpyrimidin-5-yl)imidazolidin-2-imine
SMILES COc1nc(C)nc(Cl)c1N=C2NCCN2
Specifications
Quantity 250 mg
InChI Formula InChI=1S/C9H12ClN5O/c1-5-13-7(10)6(8(14-5)16-2)15-9-11-3-4-12-9/h3-4H2,1-2H3,(H2,11,12,15)
Formula Weight 241.073
Chemical Name or Material Moxonidine