Läs mer
Tocris Bioscience™ MP A08
Beskrivning
MP A08 is a selective ATP competitive sphingosine kinase 2 and 1 (SphK2/1) inhibitor (Ki values are 6.9 and 27 μM, respectively). Exhibits selectivity for SphK 1 and 2 over other kinases. Inhibits cellular S1P production and increases sphingosine and ceramide levels in vitro. Also induces mitochondrial-associated apoptosis. Reduces tumor growth in a human lung adenocarcinoma xenograft mouse model, by inducing apoptosis and suppressing angiogenesis. Cell permeable.
Specifikationer
Specifikationer
| Kemiskt namn eller material | 4-Methyl-N-[2-[[[2-[[(4-methylphenyl)sulfonyl]amino]phenyl]imino]methyl]phenyl]benzenesulfonamide |
| CAS | 219832-49-2 |
| Synonym | NSC 122314 |
| Kvantitet | 10 mg |
| Mål | Sphingosine Kinase Inhibitors |
| Molekylformel | C27H25N3O4S2 |
| Formel vikt | 519.64 |