missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low IgG, Stem Cell Culture
Shop All MP Biomedicals Products
Click to view available options
Kvantitet:
10 g
25 g
50 g
100 g
500 g
1000 g
Unit Size:
100g
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells; IgG removed to very low levels. Lipid-rich formulation can be substituted for Albumax I™.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- IgG 0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Specifications
Specifications
| För användning med (applikation) | ELISA, Mammalian Cell Culture, Western Blot |
| Formulering | Lipid-rich |
| Molekylvikt (g/mol) | 66 |
| Namn | CellMaxx™ Bovine Albumin, Low IgG |
| pH-intervall | 6.5 to 7.5 |
| Reningsmetod | Chromatography |
| Kvantitet | 50 g |
| Källa | Bovine |
| Förvaringskrav | 4°C |
| Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction