missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Collagen from Calf Skin
Soluble form
440.96 SEK - 5697.50 SEK
Specifications
Concentration | 5mg/mL |
---|---|
Molecular Weight (g/mol) | ∽1000000 |
Name | Collagen from Calf skin |
pH Range | 3.7 |
Source | Calf Skin |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11470272
|
MP Biomedicals™
0216008450 |
50mg |
1568.80 SEK
50mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11480272
|
MP Biomedicals™
0216008480 |
100mg |
2687.10 SEK
100mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11490272
|
MP Biomedicals™
0216008483 |
250mg |
5697.50 SEK
250mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11460272
|
MP Biomedicals™
0216008410 |
10mg |
440.96 SEK
10mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Suitable for gel formation, platelet aggregation and assay of collagenase by viscometry. Prepared by method of Gallop, P.M. and Seifter, S., Meth. in Enzymol., VI: 635 (1963).
Specifications
5mg/mL | |
Collagen from Calf skin | |
Calf Skin | |
Solid |
∽1000000 | |
3.7 | |
4°C |