missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Collagen Type I from Calf Skin

Suitable for cell adhesion in tissue culture as well as gel formation, platelet aggregation and as a substrate for collagenase

Brand:  MP Biomedicals™ 0215002680

Additional Details : CAS Number : 9007-34-5 Weight : 0.00010kg

 View more versions of this product

Product Code. 11406412

  • 2729.50 SEK / 100mg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • CAS Number: 9007-34-5
  • EC Number: 232-697-4
  • MDL Number: MFCD00130825
  • Format: Powder
  • Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid

For attachment of primary cultures of epithelioid cells and many other cell types

Specifications

Specifications

Tumor Marker
100mg
Chorionic Gonadotropin from Human Pregnancy Urine
Lyophilized
3.7
Calf Skin
-20°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers