Learn More
MP Biomedicals™ Collagen Type I from Calf Skin
Suitable for cell adhesion in tissue culture as well as gel formation, platelet aggregation and as a substrate for collagenase
Brand: MP Biomedicals™ 0215002610
Additional Details : CAS Number : 9007-34-5 Weight : 0.00010kg
Description
- CAS Number: 9007-34-5
- EC Number: 232-697-4
- MDL Number: MFCD00130825
- Format: Powder
- Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid
For attachment of primary cultures of epithelioid cells and many other cell types
Specifications
Chorionic Gonadotropin from Human Pregnancy Urine | |
3.7 | |
Tumor Marker | |
10mg |
Calf Skin | |
-20°C | |
Lyophilized |