missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ CSM-HIS-TRP-URA, Powdered Media
Dropout supplement for S. cereviseae
Brand: MP Biomedicals™ 114530812
Additional Details : Weight : 0.01000kg
Description
- Triple dropout
- Complete supplement mixture of amino acid
- Complete formulations
- Dropout formulations
Formulation
- Add 0.70g of CSM-His-Trp-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined triple dropout medium
Yeast cell culture, Gene expression, Transcription regulation
Specifications
Molecular biology Certified™ | |
DOB or DOBA of 0.70g/L | |
10 g |
15°C to 30°C | |
For growth of Saccharomyces cerevisiae |