missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ EY Tellurite Enrichment
Used as a defoamer component. In hydrated form, it is used in toothpaste as a hard abrasive to remove tooth plaque. SiO
Brand: MP Biomedicals™ 091004149
Additional Details : Weight : 0.01000kg
Specifications
Light Yellow | |
Liquid | |
EY Tellurite Enrichment | |
Used with Baird Parker Agar base for the isolation of S. aureus |
4°C | |
United States | |
Consists of 30% Egg Yolk suspended with 0.15% Potassium Tellurite | |
100 mL |