Learn More
MP Biomedicals™ FastDNA™-96 Fungal and Bacterial DNA Kit, 2 x 96 Preps
FastDNA™-96 Fungal and Bacterial DNA Kit is used for rapid, high-throughput isolation of PCR-quality DNA from tough-to-lyse fungi and bacteria.
Marke: MP Biomedicals™ 119696300
Weitere Details : Weight : 1.97100kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Beschreibung
- Product Overview: The FastDNA™-96 Fungal and Bacterial DNA Kit is designed for the simple and rapid isolation of, PCR-quality DNA from tough-to-lyse fungi and bacteria in approximately 40 minutes. Gram(+/-) bacteria, fungi, including A. fumigatus, C. albicans, N. crassa, S. cerevisiae, S.pombe, and mycelium, as well as spores, nematodes, pollen, and even mammalian cells are easily lysed in approximately 60 seconds with the FastPrep-96™ Instrument.
- Applications: FastDNA™-96 Fungal/Bacterial DNA Kit is used for rapid isolation of PCR-quality DNA from tough-to-lyse fungi and bacteria in approximately 40 minutes.
- Sample Type: Spores, Bacteria (Gram +/-), Fungi, Yeast, Algae
Spezifikation
Kit | |
2 x 96 Preps | |
1 Ea. |
Spores, Bacteria (Gram +/-), Fungi, Yeast, Algae | |
FastDNA-96 Kits |
Sicherheit und Handhabung
- Danger
Gefahrenkategorie
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 C
- Specific target organ toxicity Category 3
Gefahrenhinweis
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Sicherheitshinweis
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
- MIXTURE LIST-Contains : Guanidine Thiocyanate, molecular biologyreagent
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.