Läs mer
MP Biomedicals™ Freund's Adjuvant, Complete
Produktkod. 11483966 Handla allt MP Biomedicals Produkter
Change view
Kvantitet:
50 mL
Förpackningsstorlek:
50ml
Produktkod. Kvantitet Förpackningsstorlek
11483966 50 mL 50ml
1 options
Produktkod. 11483966 Leverantör MP Biomedicals™ Leverantörsnummer 08642851
  • ONLINE EXCLUSIVE PRICE
    2135.00 SEK
    2033.07 SEK / 50ml
    Save 101.93 SEK
    4% Av
 Begränsad artikel
Beräknad leverans: 10-12-2026
Enhances the antibody response of animals injected with antigen. Made by a modification of Freund's technique
This product is made by a modification of Freund's technique, in which 25 mg of Mycobacterium is suspended in a mixture of 7.5 mL Arlacel A and 42.5 mL paraffin oil. An adjuvant is a material that when administered enhances the antibody response of animals injected with antigen.
Specifikationer
Innehåll och lagring 2°C to 8°C
För användning med (applikation) Used to enhance the antibody response of animals injected with antigen
Formulering Paraffin oil 85%, Arlacel A 15%, heat-killed Mycobacterium tuberculosis 0.05%
Kvantitet 50 mL
Produkttyp Freund's Complete Adjuvant