missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Histone from Calf Thymus

Arginine-rich subgroup

1001.70 SEK - 1749.00 SEK

Specifications

Name Histone
Source Calf
Storage Requirements 2° to 8°C
Form Powder
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11383909
View Documents Promotion Details
MP Biomedicals™
0210072625
25mg
1749.00 SEK
25mg
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
11373909
View Documents Promotion Details
MP Biomedicals™
0210072610
10mg
1001.70 SEK
10mg
View alternative products
Save Up to 
»
Estimated Shipment: 14-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Histone
Calf
2° to 8°C
Powder
Videos
SDS
Documents

Documents

Product Certifications