Learn More
MP Biomedicals™ Lipoprotein-Cholesterol Concentrate
From bovine serum
Brand:  MP Biomedicals™ 0219147680
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11464702
  • 7610.80 SEK / 100mL
Estimated Shipment: 22-05-2024
to see stock.
Description
Description
A mixture of lipoprotein cholesterol, phospholipids and fatty acids. For use as a serum-free or serum-reduced media nutrient supplement, calibration of cholesterol values, and enzymatic or nonenzymatic cholesterol determinations. Offered as a low-salt solution. No preservatives. Free of enzyme inhibitors. Filtered through 0.2μm membrane filter. Cholesterol concentration: 1000-2000mg/dL.
Specifications
Specifications
1000 to 2000mg/dL
Lipoprotein Cholesterol concentrate
100mL
0° to 5°C
Media Nutrient Supplement, Calibration of Cholesterol Values, Enzymatic or Non-enzymatic Cholesterol Determinations
7.5 to 8
Bovine Serum
Solution