missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysing Matrix F, 2 mL Tubes

Lysing Matrix F, 2 mL Tubes are used to isolate RNA and protein from samples. Lysing Matrix F is useful in applications such as isolation of intact organelles, and super-molecular structures where lysis conditions necessitate high impaction but low shearing.

Brand:  MP Biomedicals™ 116915500

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11421545

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Impact-resistant 2 mL tubes contain 1.6 mm aluminum oxide particles and 1.6 mm silicon carbide particles
  • With white caps
  • Specific Gravity: Aluminum Oxide, 3.95; Silicon Carbide, 3.20
  • Nucleic Acid Isolation and Purification
Specifications

Specifications

2 mL
Dnase-free
Lysing matrix F
500 x 2 mL Tubes
Bacteria (Gram +/-), Animal Tissues, Fungi, Plants, Coral Emulsions, Mold
Nucleic Acid isolation and purification, molecular biology, gene expression, microbiological analysis
Aluminum Oxide, Al2O3; Silicon Carbide, SiC
RNase-free
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.