Learn More
MP Biomedicals™ Papain from Papaya Latex
Sulfhydryl protease from Carica papaya latex
689.00 SEK - 13647.50 SEK
Specifications
Concentration | 2X |
---|---|
Color | Milky White |
pH | 6 to 7 |
For Use With (Application) | Produce Fab fragments of Antibodies, cell dissociation, morphological isolation of intact cortical neurons from Postnatal Rats |
Content And Storage | 4°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11344139
|
MP Biomedicals™
0210092125 |
25 mg |
689.00 SEK
25mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11364139
|
MP Biomedicals™
0210092180 |
100 mg |
1892.10 SEK
100mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11354139
|
MP Biomedicals™
0210092150 |
50 mg |
1081.20 SEK
50mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11374139
|
MP Biomedicals™
0210092190 |
500 mg |
7541.90 SEK
500mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11384139
|
MP Biomedicals™
0210092191 |
1 g |
13647.50 SEK
1g |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Papain cleaves peptide bonds of basic amino acids, leucine, or glycine. It is also hydrolyzes esters and amides.
- CAS Number: 9001-73-4
- EC Number: 232-627-2
- MDL Number: MFCD00131791
- Format: Milky white solution
- Specific Activity: ≤10 units/mg Protein
- Extinction coefficient: 25.0
- Isoelectric point (pI): pH 9.6
- Optimum pH: 6.0 to 7.0
Used to produce Fab fragments of antibodies, cell dissociation, morphological isolation of intact cortical neurons from postnatal rats. Limited papaindigestion is used for structural studies of enzymes and other proteins. In addition, papain is used in red cell serology to modify red cell surface to enhance or destroy reactivity of many red cell antigens. Useful for platelet serology studies and has also been used in enzymatic synthesis of amino acids, peptides, and other molecules.
Specifications
2X | |
6 to 7 | |
4°C | |
Papain | |
Sulfhydryl groups including heavy Metals, Carbonyl reagents, Aldehydes, Benzoylamidoacetonitrile, AEBSF, antipain, cystatin, E-64, leupeptin, PMSF, TLCK and TPCK |
Milky White | |
Produce Fab fragments of Antibodies, cell dissociation, morphological isolation of intact cortical neurons from Postnatal Rats | |
Papaya Latex | |
23000 | |
Solution |