missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Ribonuclease A, From Bovine Pancreas
Aggregates can be converted to monomers.
1033.50 SEK - 7520.70 SEK
Specifications
Concentration | ≥50 kU/mg |
---|---|
Color | White |
Purity | >70% |
pH | 7 to 7.5 |
For Use With (Application) | Used to remove RNA from DNA plasmid preparations and Protein samples |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11328659
|
MP Biomedicals™
0210107680 |
100 mg |
1033.50 SEK
100mg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11338659
|
MP Biomedicals™
0210107683 |
250 mg |
2231.30 SEK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11348659
|
MP Biomedicals™
0210107691 |
1 g |
7520.70 SEK
1g |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Ribonuclease A, [9001-99-4]
- RNase A
- From Bovine Pancreas
- Prepared from aggregate-free RNase
- Lyophilized
- Activity: 50 Kunitz units/mg.
Specifications
≥50 kU/mg | |
>70% | |
Used to remove RNA from DNA plasmid preparations and Protein samples | |
Bovine pancreas | |
13700 | |
Lyophilized Powder |
White | |
7 to 7.5 | |
0°C | |
Ribonuclease A | |
Heavy Metal ions and is competitively inhibited by DNA |