Learn More
MPP10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557135
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: DDDLQENEDN KQHKESLKRV TFALPDDAET EDTGVLNVKK NSDEVKSSFE KRQEKMNEKI ASLEKELLEK KPWQLQGEVT AQKRPENSLL EETLHFDHAV R Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 83%.
MPP10 Polyclonal antibody specifically detects MPP10 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
MPP10 | |
Polyclonal | |
Unconjugated | |
MPHOSPH10 | |
2810453H10Rik; 5730405D16Rik; AI326008; CT90; M phase phosphoprotein 10; M-phase phosphoprotein 10; M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein); Mphosph10; MPP10; MPP10P; PPP1R106; protein phosphatase 1, regulatory subunit 106; U3 small nucleolar ribonucleoprotein; U3 small nucleolar ribonucleoprotein protein MPP10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10199 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00566 | |
MPHOSPH10 | |
Recombinant protein corresponding to Human MPP10 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only